Sarcosine sodium salt ∼35% in water
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
A3280-100ML
A3280-25ML
TCIAA3280-100MLEA
846
DKK
TCIAA3280-100ML
TCIAA3280-25ML
Sarcosine sodium salt ∼35% in water
Sarcosine sodium salt
Formel:
C₃H₇NO₂ |
MDL Number:
MFCD00068282 CAS-nummer: 4316-73-8 UN nummer: 3267 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...