Brady® Paper Label Tape for BradyJet™ J7300 Colour Label Printer
Leverandør: Brady
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
176588 176587 170471
BRDY176588EA 1360 DKK
BRDY176588 BRDY176587 BRDY170471
Brady® Paper Label Tape for BradyJet™ J7300 Colour Label Printer
Etiketter
Low cost and high contrast, these inkjet receptive paper labels offer excellent smear resistance for economical light-duty identification.
  • Permanent acrylic adhesive ensures labels adhere strongly to a variety of flat and curved surfaces
  • Inkjet receptive paper material
  • Designed for indoor use
Ideal for barcoding, quality and control (testing/production, circle labels, date/time), and warehousing racks, bins, and storage. Permanent acrylic adhesive ensures labels adhere strongly to a variety of flat and curved surfaces, including cardboard, plastic and metal. Built-in LabelSense™ technology enables cartridge and printer communication for fast changeovers, automatic label setup, and seamless software interaction.
For use with the BradyJet™ J7300 Colour Label Printer. Ink cartridges are required to print on this material; use part numbers P-CMY-1 and P-K-1.