Magnetic Polyester Labels B-2509 for BradyJet™ J7300 Inkjet Printer
Leverandør: Brady
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
170451 170450
BRDY170451EA 1830 DKK
BRDY170451 BRDY170450
Magnetic Polyester Labels B-2509 for BradyJet™ J7300 Inkjet Printer
Etiketter
Experience effortless removal and repositioning with magnetic labels for the BradyJet™ J7300 Printer. Get the flexibility needed without adhesive residue or surface damage. For use on smooth surfaces.
  • Ideal for reusable, repositionable, and Lean/5S identification applications such as warehousing racks, bins, and storage
  • Magnetic polyester construction
  • Designed for indoor use
  • Abrasion-resistant, removable, solvent/chemical resistant
  • Matt finish
Magnetic labels facilitate visual workplace best practices, improve efficiency, and reduce errors in a wide variety of settings. Magnetic labels withstand moderate heat, mild chemicals, oil, and water. Easiest and quickest labelling option when removal and repositioning are frequent. Built-in LabelSense™ technology enables cartridge and printer communication for fast changeovers, automatic label setup, and seamless software interaction.
For use with the BradyJet™ J7300 colour label printer. Ink cartridges are required to print this material; use part numbers P-CMY-1 and P-K-1.
Certificeringer: 2011/65/EU and 2015/863