Showa DURACoil 546W Cut Resistant Gloves
Leverandør: Showa
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
546W-10/XXL 546W-6/S 546W-7/M 546W-8/L 546W-9/XL
BEST546W-10/XXLEA 1240 DKK
BEST546W-10/XXL BEST546W-6/S BEST546W-7/M BEST546W-8/L BEST546W-9/XL
Showa DURACoil 546W Cut Resistant Gloves
Handsker
Featuring an engineered DURACoil liner and reinforced with HPPE, this cut- and abrasion-resistant model is an ultra-comfortable, multi-use work glove.
  • Cut-resistant, abrasion-resistant and oil-resistant
  • Seamless knit
  • Flexible and breathable
  • Form-fitting
  • Increased dexterity
  • Machine washable
The foamed polyurethane palm coating provides good grip and breathability, and the light color makes it easy to identify soiling and contamination. A seamless and ergonomic design replicates the natural curves of the hand for enhanced dexterity and comfort over long periods of wear.
Certificeringer: Cat II. EN 388:2016+A1:2018, 4X42C.