Showa 545 Cut Resistant Gloves, Nitrile Palm Coated
Leverandør: Showa
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
545-8/L 545-9/XL 545-6/S 545-7/M
BEST545-8/LEA 1950 DKK
BEST545-8/L BEST545-9/XL BEST545-6/S BEST545-7/M
Showa 545 Cut Resistant Gloves, Nitrile Palm Coated
Handsker
The nitrile technology of the durable Showa 545 gloves ensure superb grip as well as oil and cut resistance.
  • Cut-resistant
  • Oil-resistant
  • Seamless knit
  • Extra grip
  • Flexible
  • Breathable
When working outdoors or in dirty environments, the elasticated wrist cuff prevents debris from entering the glove to ensure comfort over long periods of wear. Ensure precision and dexterity, even when assembling greasy components, with the 545 HPPE gloves.
Certificeringer: Cat II. EN 388:2016+A1:2018, 4242B.