Showa 406 Cold Weather Gloves
Leverandør: Showa
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
406-10/XXL 406-9/XL 406-7/M 406-8/L
BEST406-10/XXLEA 10100 DKK
BEST406-10/XXL BEST406-9/XL BEST406-7/M BEST406-8/L
Showa 406 Cold Weather Gloves
Handsker
Designed to protect hands from liquid and freezing temperatures, Showa 406 water-repellent natural rubber latex gloves allow for optimum comfort when working in tough conditions.
  • Water-resistant and abrasion-resistant
  • Increased visibility
  • Protects down to –30 °C
  • Breathable
  • Thermal insulation
Intelligent insulation keeps hands warm and dry while reducing perspiration inside the glove, to preserve grip and tactility. Even when wet, the glove's rough surface allows for increased control, and the flexible material enables easy movement.
Certificeringer: Cat II. EN 388:2016+A1:2018, 2131X. EN 407:2020, X2XXXX. EN 511:2006 010.
Emballering: Gloves are packaged 5 pair per polybag, 12 polybags per case (60 total pair per case).