Showa 257X Cut Resistant Gloves, Uncoated
Leverandør: Showa
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
257X-10/XXL 257X-6/S 257X-7/M 257X-8/L 257X-9/XL
BEST257X-10/XXLEA 1730 DKK
BEST257X-10/XXL BEST257X-6/S BEST257X-7/M BEST257X-8/L BEST257X-9/XL
Showa 257X Cut Resistant Gloves, Uncoated
Handsker
This cut level A7 aramid glove is designed specifically for use in automotive, stamping, and industrial applications.
  • Cut-resistant and abrasion-resistant
  • Increased dexterity
  • Lightweight and breathable
  • Machine washable
  • Form-fitting
  • Touch screen compatible
A 13 gauge lightweight design offers high dexterity, very high cut resistance, and can be laundered multiple times without compromising the glove’s performance. Thanks to the lightweight and thin construction, the 257X makes a great liner to additional workwear or PPE.
Certificeringer: Cat II. EN 388:2016+A1:2018, 4X42F. EN 407:2020, X1XXXX.
Emballering: Gloves are packaged 12 pair per polybag, 6 polybags per case (72 total pair per case).