Showa 234 Cut Resistant Gloves, Foam Nitrile Dipped
Leverandør: Showa
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
234-10/XXL 234-6/S 234-7/M 234-8/L 234-9/XL
BEST234-10/XXLEA 1530 DKK
BEST234-10/XXL BEST234-6/S BEST234-7/M BEST234-8/L BEST234-9/XL
Showa 234 Cut Resistant Gloves, Foam Nitrile Dipped
Handsker
Designed for use in 'knife hand' applications in food processing.
  • Cut-resistant and abrasion-resistant
  • Flexible and breathable
  • Lightweight and increased dexterity
  • Machine washable
  • Seamless knit
  • Heat-resistant and hydrocarbon-resistant
  • Form-fitting
This flexible cut protection glove combines foam nitrile with a HPPE reinforced liner to provide excellent cut resistance performance. A foam nitrile coating protects against oils, hydrocarbons, grease, and abrasions, while offering excellent grip in wet and dry conditions. Cooling properties of HPPE and a breathable design reduce perspiration to keep hands dry.
Certificeringer: Cat II. EN 388:2016+A1:2018, 4X42D. EN 407:2020, X1XXXX. Food contact approved.
Emballering: Gloves are packaged 12 pair per polybag, 6 polybags per case (72 total pair per case).