Labels B-427 Small Core 25 mm Thermal Transfer Printers
Leverandør: Brady
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
236987 800538 236986 236989 217096 217094 236988 236998
BRDY236987EA 3290 DKK
BRDY236987 BRDY800538 BRDY236986 BRDY236989 BRDY217096 BRDY217094 BRDY236988 BRDY236998
Labels B-427 Small Core 25 mm Thermal Transfer Printers
Etiketter
Printable, self-laminating vinyl labels (B-427) for thermal transfer printers.
  • Self-extinguishing
  • Excellent water and oil resistance
  • Excellent abrasion and smudge resistance
Certificeringer: This material is UL recognised with its respective ribbon.