Chemical Resistant Polyester Slide Labels StainerBondz™ B-481
Leverandør: Brady
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
170885 170886
BRDY170885EA 447 DKK
BRDY170885 BRDY170886
Chemical Resistant Polyester Slide Labels StainerBondz™ B-481
Etiketter
Easily create chemical-resistant microscope slide labels with the StainerBondz™ polyester label material (B-481). Print almost anywhere with a Brady handheld label printer.
  • Specialised - Available in presized shapes for commonly used slides and tissue cassettes in which superior chemical resistance is required
  • Durable - Withstands the rigors of typical deparaffinisation including ethanol and xylene, as well as hematoxylin and eosin slide staining processes
  • Adhesive - Made with a permanent acrylic adhesive for tight bonding onto smooth surfaces in extreme