Showa 542X Cut Resistant Gloves
Leverandør: Showa
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
542X-6 542X-7 542X-8 542X-9
BEST542X-6EA 1250 DKK
BEST542X-6 BEST542X-7 BEST542X-8 BEST542X-9
Showa 542X Cut Resistant Gloves
Handsker
These cut and abrasion resistant gloves made from HPPE protect hands during component assembly and hazardous applications.
  • Seamless fabrication
  • Flexible, tight fit
  • Breathable
  • Abrasion resistant/cut resistant
  • Uncoated finish
  • HPPE reinforced
  • Elastic cuff/ergonomic
  • Rough grip surface
The 542 uses HPPE to provide high cut and abrasion resistance while maintaining maximum flexibility and dexterity. An open back reduces perspiration and the seamless finish reduces skin irritation during extended wear. Applications are white goods, quality control.
Certificeringer: EN 420:2003 + A1:2009 Cat. II, En 388 (2.2.4.0.A).
Emballering: Pair per polybag: 10; Polybag per box: 20; Pair per box: 200.