Transfectamine™ mRNA Transfection Reagent
Leverandør: AAT BIOQUEST
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
60029 60031 60030
AATB60029EA 406 DKK
AATB60029 AATB60031 AATB60030
Transfectamine™ mRNA Transfection Reagent
Transfektionsreagenser
Transfectamine™ mRNA Transfection reagent is a powerful and versatile transfection reagent designed to introduce a higher amount of mRNA into eukaryotic cells, or more specifically, into animal cells.
  • Consistently high transfection efficiency
  • Higher viability of transfected cells
  • Does not require special medium
Transfectamine™ mRNA Transfection reagent does not require special medium and is easier to use compared to most of the commercial transfection reagents.
Advarsel: For research use only.