Transfectamine™ 6000 CRISPR Transfection Reagent
Leverandør: AAT BIOQUEST
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
60023 60024 60028
AATB60023EA 3510 DKK
AATB60023 AATB60024 AATB60028
Transfectamine™ 6000 CRISPR Transfection Reagent
Transfektionsreagenser
Transfectamine™ 6000 CRISPR Transfection reagent is a non-liposomal formula designed for optimal delivery and effectiveness of CRISPR/Cas9 components across a wide array of cell types.
  • Consistently high transfection efficiency
  • Higher viability of transfected cells
  • Does not require special medium
Transfectamine™ 6000 CRISPR Transfection reagent is suitable for various applications, such as gene knockout and knock-in experiments, the generation of CRISPR-mediated gene-edited cell lines, functional genomics studies, and research in therapeutic gene editing.
Advarsel: For research use only.