Removable Paper Labels B-403 for Thermal Printing
Leverandør: Brady
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
149608 152248
BRDY149608EA 270 DKK
BRDY149608 BRDY152248
Removable Paper Labels B-403 for Thermal Printing
Etiketter
For the ultimate solution in label and adhesive removal, choose Brady’s B-403 Water Dissolvable Label. Ideal for laboratories, temporary identification needs, work-in-process product flows, and other general identification applications.
  • Matt finish
  • Work-In-Process - warm water and 30 seconds is all it takes to completely remove the label and adhesive
  • Application versatility: From laboratories to restaurants, this dissolvable paper material is an excellent solution for temporary identification of any kind
  • Black print on white paper with easy to apply pressure-sensitive adhesive
For printers BMP21-LAB, BMP21-PLUS, IDPAL, LABPAL, M210, M210-LAB, M211
Component and equipment labelling, electrical labelling, facility identification, floor marking, laboratory labelling, lean and 5S labelling, pipe and valve marking, warehouse marking.
Certificeringer: RoHS compliant to 2011/65/EU (Pass 2015/863