Reversible Biotin succinimidyl ester, ReadiLeave™
Leverandør: AAT BIOQUEST
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
3401 3400
AATB3401EA 3000 DKK
AATB3401 AATB3400
Reversible Biotin succinimidyl ester, ReadiLeave™
Reversible Biotin succinimidyl ester
ReadiLeave™ Reversible (RLR) Biotin is a newly developed biotin derivative that has significantly reduced affinity to avidin (including streptavidin) to make the binding of RLR biotin and streptavidin readily reversible when needed.
  • Stable for 12 months
Quality tested in-house
Advarsel: For Research Use Only