Psoralen-PEG3-Biotin
Leverandør: AAT BIOQUEST
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
39050 39051
AATB39050EA 993 DKK
AATB39050 AATB39051
Psoralen-PEG3-Biotin
Psoralen-PEG3-Biotin
Psoralen-PEG3-Biotin is a photoactivatable reagent for biotinylating DNA or RNA.
  • Stable for 24 months
Quality tested in-house
Advarsel: For Research Use Only