Beta Sterile Kits
Leverandør: ISOFIELD IRELAND
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
80451S 80451M 80451L 80451XL 80451XXL 80451XXXL
ISOF80451SEA 8430 DKK
ISOF80451S ISOF80451M ISOF80451L ISOF80451XL ISOF80451XXL ISOF80451XXXL
Beta Sterile Kits
Engangsbeklædning
Sterile coverall with mandarin collar, hood with integrated facemask, and overboots.
  • Manufactured in a facility operating under ISO 9001:2015 quality management system
  • Processed in a NEBB certified ISO Class 5 cleanroom
Mandarin collar coverall:
GMP Grade A and B, ISO Class 4 and 5 cleanroom compatible
Two-layer mandarin collar provides secure fit at neck area
Zip flap with double-sided tape prevents ingress and egress of particles
Low-linting, anti-static fabric suitable for chemo handling
Elasticated thumb loops
Back elastic to prevent bellows effect
Hood with integrated facemask:
Integrated mask with ties for quick and easy donning
Latex-free enclosed elastic face opening for comfortable fit
Extra-long yoke for added protection
Three panel hood design to minimise skin exposure
Overboots:
Extra-long knee height
Two-ties for added security
Anti-slip PU sole
Emballering: 2 sets (hood/coverall/boots) per sealed inner bag. 1 inner bag per sealed outer bag. 40 outer bags per carton liner with twist tie (S to XXL). 35 sets per carton (3XL).