Anti-CD79a Mouse Monoclonal Antibody [clone: ABT-CD79a]

Leverandør: ANTIBODIES.COM
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

A99690-100 A99690-50
ANTIA99690-100EA 3550 DKK
ANTIA99690-100 ANTIA99690-50
Anti-CD79a Mouse Monoclonal Antibody [clone: ABT-CD79a]
Antistoffer
Mouse monoclonal (ABT-CD79a) antibody to CD79a for IHC with samples derived from human.

  • Validated Applications: IHC

Type: Primary
Antigen: CD79a
Clonality: Monoclonal
Clone: ABT-CD79a
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR