Anti-GAPDH Mouse Monoclonal Antibody [clone: GA1R]

Leverandør: ANTIBODIES.COM
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

A85271-100 A85271-200
ANTIA85271-100EA 2650 DKK
ANTIA85271-100 ANTIA85271-200
Anti-GAPDH Mouse Monoclonal Antibody [clone: GA1R]
Antistoffer
Mouse monoclonal (GA1R) antibody to GAPDH for Dot, ELISA, IS and WB with samples derived from BL-21 Bacteria, chicken, Hamster, human, mouse, rat, rabbit, Saccharomyces cerevisiae and Sf9 Insect.

  • Validated applications: Dot, ELISA, IS, W

Type: Primary
Antigen: GAPDH
Clonality: Monoclonal
Clone: GA1R
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: BL-21 Bacteria;Chicken;Hamster;Human;Mouse;Rat;Rabbit;Saccharomyces cerevisiae;Sf9 Insect
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR