Anti-SOX9 Mouse Monoclonal Antibody [clone: ABT-SOX9]
Leverandør: ANTIBODIES.COM
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
A99228-100 A99228-50
ANTIA99228-100EA 3550 DKK
ANTIA99228-100 ANTIA99228-50
Anti-SOX9 Mouse Monoclonal Antibody [clone: ABT-SOX9]
Antistoffer
Mouse monoclonal (ABT-SOX9) antibody to SOX9 for IHC and WB with samples derived from human.
  • Validated applications: IHC, WB
Type: Primary
Antigen: SOX9
Clonality: Monoclonal
Clone: ABT-SOX9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human