Anti-Prolactin Mouse Monoclonal Antibody [clone: ABT-PRL]
Leverandør: ANTIBODIES.COM
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
A98910-100 A98910-50
ANTIA98910-100EA 3550 DKK
ANTIA98910-100 ANTIA98910-50
Anti-Prolactin Mouse Monoclonal Antibody [clone: ABT-PRL]
Antistoffer
Mouse monoclonal (ABT-PRL) antibody to Prolactin for IHC with samples derived from human.
  • Validated applications: IHC
Type: Primary
Antigen: Prolactin
Clonality: Monoclonal
Clone: ABT-PRL
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human