Anti-MyoD1 Mouse Monoclonal Antibody [clone: ABT-MYOD1]
Leverandør: ANTIBODIES.COM
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
A98670-100 A98670-50
ANTIA98670-100EA 3550 DKK
ANTIA98670-100 ANTIA98670-50
Anti-MyoD1 Mouse Monoclonal Antibody [clone: ABT-MYOD1]
Antistoffer
Mouse monoclonal (ABT-MYOD1) antibody to MyoD1 for IHC and WB with samples derived from human.
  • Validated applications: IHC, WB
Type: Primary
Antigen: MyoD1
Clonality: Monoclonal
Clone: ABT-MYOD1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human