Anti-ErbB 2 Mouse Monoclonal Antibody [clone: ABT-Her2]

Leverandør: ANTIBODIES.COM
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

A99844-100 A99844-50
ANTIA99844-100EA 3550 DKK
ANTIA99844-100 ANTIA99844-50
Anti-ErbB 2 Mouse Monoclonal Antibody [clone: ABT-Her2]
Antistoffer
Mouse monoclonal (ABT-Her2) antibody to Her 2 for IHC with samples derived from Human.

  • Validated Applications: IHC

Type: Primary
Antigen: ErbB 2
Clonality: Monoclonal
Clone: ABT-Her2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR