3'-O-Azidomethyl-dGTP
Leverandør: AAT BIOQUEST
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
17224 17225
AATB17224EA 5030 DKK
AATB17224 AATB17225
3'-O-Azidomethyl-dGTP
Nukleinsyrereagenser Nukleotider
3'-O-Azidomethyl-dGTP is one of the four essential dNTPs (A, C, G, T) that can be used for the DNA sequencing by synthesis (SBS).
  • Highly purified
DNA sequencing is a fundamental tool in biological and medical research (especially for personalised medicine).
Advarsel: For research use only.