Anti-GCSF Receptor Rabbit Recombinant Antibody [clone: DM172]

Leverandør: ANTIBODIES.COM
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

A318537-100 A318537-500
ANTIA318537-100EA 4780 DKK
ANTIA318537-100 ANTIA318537-500
Anti-GCSF Receptor Rabbit Recombinant Antibody [clone: DM172]
Antistoffer
Recombinant rabbit monoclonal [DM172] antibody to GCSF Receptor for ELISA and Flow cytometry with samples derived from human.

  • Validated applications: ELISA, Flow cytometry
  • Recommended dilutions: ELISA: 1:5000 to 10000, Flow cytometry: 1:100

Type: Primary
Antigen: GCSF Receptor
Clonality: Recombinant
Clone: DM172
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR