Anti-IgG Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))

Leverandør: AAT BIOQUEST
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

50260 50261
AATB50260EA 896 DKK
AATB50260 AATB50261
Anti-IgG Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antistoffer
Anti-human secondary antibodies have well-characterised specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target.

  • Affinity purified
  • Easily reconstituted

Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence).

Advarsel: For research use only.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Goat
Isotype: IgG (H, L)
Reactivity: Human
New Product
Bestil nu

Specifikationer

Antigen symbol IgG
Environmentally Preferable
Antigen name Immunoglobulin G
Antistof type Secondary
Clonality Polyclonal
Konjugation HRP (Horseradish Peroxidase)
Reactivity Human
Vært Goat
Isotype IgG (H, L)
Western blot Yes
ELISA Yes
ImmunoChemistry Yes
Flowcytometri Yes
Krydsadsorption No
Opbevaringstemperatur Freeze (<–15 °C); Minimise light exposure
Shipping temperature 20

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR