Anti-IgG Goat Polyclonal Antibody *Cross Adsorbed* (iFluor® 790)

Leverandør: AAT BIOQUEST
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

50126 50127
AATB50126EA 1150 DKK
AATB50126 AATB50127
Anti-IgG Goat Polyclonal Antibody *Cross Adsorbed* (iFluor® 790)
Antistoffer
Anti-human secondary antibodies have well-characterised specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target.

  • Affinity purified
  • Easily reconstituted

Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence).

Advarsel: For research use only.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: iFluor® 790
Epitope:
Host: Goat
Isotype: IgG (H, L)
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR