Meliseptol® Acute Alcoholic Surface Disinfectant Kit
Leverandør: B. Braun
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
180243 180255
BRAU180243EA 394 DKK
BRAU180243 BRAU180255
Meliseptol® Acute Alcoholic Surface Disinfectant Kit
Renseopløsninger Disinfektanter
Fast acting alcohol based disinfectant for small patient near surfaces and medical devices – effective against bacteria, yeasts, mycobacteria, enveloped and non-enveloped viruses within a minute.
  • Solution for rapid disinfection of non-invasive medical devices and small areas
  • Ready-to-use alcohol based disinfectant
  • Broad efficacy: Full virucidal activity
  • Good material compatibility
  • Perfume-free, mild smell
Meliseptol® acute is a fast and effective ready-to-use alcoholic surface disinfectant, ideal for alcohol-resistant materials. It offers a short exposure time and is free from quaternary ammonium compounds (QAC), aldehydes, and alkylamines, ensuring a safer application. This colorless disinfectant is conveniently available in wipes packaged in Flowpacks, making it easy to use. Moreover, it is listed on both the VAH list and the IHO List of Disinfectants, emphasizing its reliability and compliance with industry standards.
Compatible with B.Braun wipes dispenser system.
Pour Meliseptol® acute from the dosing bottle onto a disposable cloth and wet the surfaces completely. Or spray the surface to be treated with the spray bottle until it is completely wet and rub the disinfectant with a cloth. The surfaces must not be rubbed dry. Materials that are sensitive to alcohol and acids (e.g. acrylic glass) should be tested beforehand for their resistance to Meliseptol® acute.
Use disinfectants safely. Always read the label and product information before use.