Stainless Steel Static Wire Modular Shelving Units
Leverandør: TEKNOMEK
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
SHSH-MSWI10X60 SHSH-MSWI25X60
TEKMSHSH-MSWI10X60EA 0 DKK
TEKMSHSH-MSWI10X60 TEKMSHSH-MSWI25X60
Stainless Steel Static Wire Modular Shelving Units
Møbler Hylder
Stainless steel modular shelving units with removeable wire shelves are hygienic, ingress-free and manufactured in 304-grade stainless steel. All shelves are designed so you can just remove an individual shelf for cleaning.
  • 600 kg payload per unit
  • Adjustable feet on static versions
  • Units over 1500 mm feature 3 uprights (1 in the centre)
  • Comes with four or six plastic floor levelling adjustable feet
Each unit bay is supplied with 4 shelves as standard but each unit can hold up to 10 shelves, ensuring that you will find a unit suitable for your storage needs.
Support points every 150 mm make the shelving positions very flexible. You can even build a corner unit using hooks (2 per shelf), without any uprights needed. Units over 1500 mm feature 3 uprights (1 in the centre).
The maximum payload per unit is 600 kg (225 kg mobile). With individual shelf loading for under 1000 mm width: 150 kg and over 1000 mm width: 100 kg.
It comes with floor levelling adjustable feet to help compensate for any unevenness in the work area. Stainless steel castors are available but they are designed to facilitate the movement of the shelving units for cleaning purposes, rather than for transportation.