Foaming Solutions for Hygienic Hand Washing
Leverandør: PHAGOGENE
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
65123 SA0141050
PHAG65123EA 523 DKK
PHAG65123 PHAGSA0141050
Foaming Solutions for Hygienic Hand Washing
Hudpleje Sæber og huden rensemidler
Liquid washing and disinfecting soap specially formulated for the hygienic hands washing.
  • Skin-friendly formula even in case of frequent washing
  • Leave your hands soft
  • Enriched with glycerine
  • Without perfume
Used before any aseptic gesture (catheters and more) and systematically before and after any contact with a patient or contaminating material. Specially formulated for the medical, food and institutional sectors.
Appearance: Gelled liquid, opalescent
Colour: Colourless
Odor: Alcohol
pH at 20 °C (pure product): 7,0 ±0,5
Density at 20 °C (g/ml): 0,905 ±0,005
Advarsel: Use biocides safely. Before use, read the label and product information.