Anti-SHP2 (NT) Rabbit Polyclonal Antibody
Leverandør: ENZO LIFE SCIENCES
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
PSC-3903-C020 PSC-3903-C100
ENZOPSC-3903-C020EA 0 DKK
ENZOPSC-3903-C020 ENZOPSC-3903-C100
Anti-SHP2 (NT) Rabbit Polyclonal Antibody
Antistoffer
Advarsel: For research use only.
Type:
Antigen: SHP2 (NT)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat