Single Use Oral Syringes, 3-Parts
Leverandør: CODAN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
62.4960 62.4980 62.6960
CODA62.4960EA 0 DKK
CODA62.4960 CODA62.4980 CODA62.6960
Single Use Oral Syringes, 3-Parts
Sprøjter Sprøjter, medicinske
The non-sterile oral syringe program consist of reliable, precise, high-quality single use syringes, includes a range with oral tip dispensers with clear and amber barrel for UV light protection of sensitive medications.
  • Three-part syringe with thin silicone ring
  • Easy and very even withdrawal and emptying
  • Smooth plunger guidance
  • Reliable plunger stop
  • Precise scale with dual scale printing
  • Environmentally friendly and PVC-free materials
  • Latex-free
  • Balanced portfolio with centric and eccentric connections
The oral syringes also meets the highest standards for feeding premature infants when breast milk has to be administered via an oral syringe and provide support for sensitive handling. This applies also to all other areas in which oral syringes are used.
The amber coloured oral syringes provide optimal light protection, allowing safe oral administration of light-sensitive medications.
These oral syringes have an extra wide oral cone that makes the syringe distinctive and prevents accidental connection to an administration system.