Anaerobic systems, workstation, Whitley A35
Leverandør: DON WHITLEY SCIENTIFIC
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
A07000 A07100
710-0842EA 417540 DKK
710-0842 710-0843
Anaerobic systems, workstation, Whitley A35
Anaerobe systemer
Sample manipulation within a secure anaerobic environment is now as easy as working aerobically at the laboratory bench.
  • Rapid, built-in airlock with mechanical and electrical interlocks – no risk of compromising conditions inside the chamber; The airlock is flushed with nitrogen as an additional cost saving
  • Innovative colour touch screen interface with PIN code-protected user levels
  • Available with the Instant Access Porthole System that allows rapid access to the chamber or with the unique oval, sleeved ports
  • Dual gas operation and automatic commissioning cycle as standard
  • Automatic dehumidifier – no user maintenance necessary
  • Both the working and storage areas of the chamber are temperature controlled, allowing the full internal volume to be used for incubation
  • Bright, even illumination ensures sufficient light for any tasks performed inside the chamber
  • Available with HEPA Filtration System that ensures levels of atmospheric cleanliness inside the workstation that exceed the requirements of ISO 14644 Class 3
The Whitley A35 Anaerobic Workstation fulfils the diverse specifications of both research and clinical customers. This workstation accommodates up to 600 x 90mm Petri dishes.
Note: The cycle time mentioned in the specifications is for the airlock.
Bestillingsoplysninger: Options and accessories not included (sold separately). An optional Anaerobic Conditions Monitor provides confirmation that anaerobic conditions exist inside the workstation and provides an early indication if optimal growth conditions begin to vary. For this and other available options and accessories, please see the attached brochures and contact your Avantor representative.