VWR®, kompakte vakuumpumper til filtrering og fastfaseudsugning
Leverandør: VACUUBRAND
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
721111 721112 20721010 721011 721012 20721110
181-0239EA 8040 DKK
181-0239 181-0240 181-0235 181-0236 181-0237 181-0238
VWR®, kompakte vakuumpumper til filtrering og fastfaseudsugning
Pumper Membranpumper, vakuum
Vacuum filtration is one of the most common applications used for sample preparation in chemistry, microbiology, wastewater control and other analytical processes. VP 100 and VP 100 C diaphragm pumps offer a compact, high performance and easy to use solution which is perfect for both single and multiple filtrations.
  • Convenient, quick and simple to use due to the top-mounted power switch
  • Whisper-quiet with ultra-low vibration resulting from a patented motor mounting
  • Requires minimal bench space
  • Maintenance-free drive system and proven long diaphragm life
  • High chemical resistance (especially VP 100 C)
Based on the highly successful technology used in the three-stage diaphragm pump series MD 1 and MD 1C, the VP 100 provides a well proven and extraordinarily long diaphragm lifetime. The functional, space saving, innovative design, with visible top-mounted power switch, ensures convenient and quick operation for day-to-day laboratory work. Robust PTFE diaphragms and valves provide optimum chemical resistance. An optional manual control valve with gauge enables variable fine adjustment of the pumping speed.