Butyl (S)-(-)-lactat ≥97%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
A13821.0B
A13821.0I
A13821.30
A13821.0BEA
1280
DKK
A13821.0B
A13821.0I
A13821.30
Butyl (S)-(-)-lactat ≥97%
Butyl (S)-(-)-lactat
Purity: 97+%
CAS No.: 34451-19-9
Molecular Formula: C7H14O3
Formula Weight: 146.19
Flash Point: 69°(157°F)
Refractive Index: 1.4210
Boiling Point: 185-187°
MDL No.: MFCD00004519
Formel:
C₇H₁₄O₃ Kogepunkt: 185…187 °C Smeltepunkt: < 0 °C Massefylde: 0,984 Flammepunkt: 69 °C (157 °F) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00004519 CAS-nummer: 34451-19-9 EINECS: 252-036-3 |