KingFisher™ Deep Well Plates, Barcoded
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
95040450B 95040470B 97002524B 97002534B 97002540B 97002610B 95040460B 95040480B 97002620B 97002820B
FINN95040450BEA 2250 DKK
FINN95040450B FINN95040470B FINN97002524B FINN97002534B FINN97002540B FINN97002610B FINN95040460B FINN95040480B FINN97002620B FINN97002820B
KingFisher™ Deep Well Plates, Barcoded
Mikroplader Deepwell Mikroplader
Optimise performance using Thermo Scientific™ KingFisher™ barcoded plastics designed specifically for use with the KingFisher Flex, Duo Prime, Presto and Apex instruments.
  • KingFisher instruments utilise disposable plastic plates and tip combs made of polypropylene
  • Ideal for magnetic particle processing due to their low binding affinity for biomolecules
Both the yield and quality of the isolated protein or nucleic acid are significantly improved with special plates and tip combs designed for KingFisher systems.