Multipet Pro 8-Channel Micropipettes
Leverandør: KARTELL
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
0170303 0170403 0170503 0170603 0170703 0170803
KART0170303EA 5750 DKK
KART0170303 KART0170403 KART0170503 KART0170603 KART0170703 KART0170803
Multipet Pro 8-Channel Micropipettes
Pipetter Automatiske pipetter
μAir™ technology ensures high precision, exceeding ISO 8655 standards, by minimizing dead air space between the piston and liquid in the tip. This technology reduces the air compression and allows the user to achieve greater precision during laboratory practices.
  • Totally autoclavable at 121 °C for 20 minutes
  • Patented UniCal® technology facilitates quick recalibration in a single operation
  • Ultra soft plunger force greatly improves user comfort throughout extended use
  • Smooth outer surface allows easy cleaning
  • Premium aesthetics and build quality
  • Ultra lightweight pipette (PC)
  • Universal tip cones are compatible with most internationally accepted standard tips
The multichannel pipette is also recommended for ELISA protocols, molecular screening, kinetic studies, DNA amplification, etc. The head can be rotated 360° to ensure flexible use. The user can thus select the rotation angle that best suits the layout of the workstation for comfortable operation.
Patented UniCal® technology facilitates a less than 15 second quick recalibration in a single operation, without disengaging the digits from the plunger mechanism and avoiding traditional calibration methods. Ultra soft plunger force, with its first step plunger force being only 8,95 N, and the second step plunger force being only 16,85 N. The soft plunger greatly improves user comfort even during long hours of pipetting.
Certificeringer: Individually calibrated in an ISO 17025 accredited laboratory according to ISO 8655 standards.
Emballering: Supplied in single carton with calibration certificate included.