ESD Occupational Shoes, Lacing Sytem, Riley XXF Boa® GTX Low O2 WR CI
Leverandør: ELTEN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
972670/41 972670/42 972670/43 972670/44 972670/45 972670/46 972670/47 972670/35 972670/36 972670/37 972670/38 972670/39 972670/40 972670/48
ELTN972670/41EA 858 DKK
ELTN972670/41 ELTN972670/42 ELTN972670/43 ELTN972670/44 ELTN972670/45 ELTN972670/46 ELTN972670/47 ELTN972670/35 ELTN972670/36 ELTN972670/37 ELTN972670/38 ELTN972670/39 ELTN972670/40 ELTN972670/48
ESD Occupational Shoes, Lacing Sytem, Riley XXF Boa® GTX Low O2 WR CI
Sko og støvler
ESD work shoe made from hydrophobized suede and hydrophobized textile upper and a PU/PU sole (Wellmaxx Flex).
  • Full, padded bellows tongue
  • All weather Gore-Tex performance membrane
  • Boa® fit lacing system
  • Full-length inlay sole ESD PRO
  • Soft fleece ESD insole
Suitable for use Indoors and outdoors, areas where exposure to moisture is expected (O2), a risk of electrostatic discharge (ESDS/ESD), areas where there is no risk of falling heavy objects.
Colour: Blue/lime
Certificeringer: EN ISO 20347 O2 FO/SR/WR/CI, form A.