ESD Safety Ankle Boots, Lacing System, Matthew Pro Boa® GTX Mid S3 CI Type 1
Leverandør: ELTEN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
1768501/38 1768501/39 1768501/36 1768501/37 7685501/40 7685501/41 7685501/42 7685501/43 7685501/44 7685501/45 7685501/46 7685501/47 7685501/48
ELTN1768501/38EA 1050 DKK
ELTN1768501/38 ELTN1768501/39 ELTN1768501/36 ELTN1768501/37 ELTN7685501/40 ELTN7685501/41 ELTN7685501/42 ELTN7685501/43 ELTN7685501/44 ELTN7685501/45 ELTN7685501/46 ELTN7685501/47 ELTN7685501/48
ESD Safety Ankle Boots, Lacing System, Matthew Pro Boa® GTX Mid S3 CI Type 1
Sko og støvler
ESD safety ankle boot made from hydrophobised microfibre/hydrophobised textile Cordura® upper and a PU/PU sole (Ergo-Active 2). Type 1: For average wide feet with long toes, medium ball and heel width, flat ball angle.
  • Gore-Tex Crosstech® membrane
  • Full, padded bellows tongue
  • Semi-orthopaedic inlay sole ESD black
  • Boa® fit lacing system
  • Non-metallic, penetration-resistant midsole
  • Steel toe cap
  • TPU scuff cap
Suitable for indoors and outdoors, areas with exposure to moisture (S2), risk of electrostatic discharge (ESDS/ESD) and areas where there is risk of penetration from pointed and sharp objects (S3).
Colour: Black
Certificeringer: EN ISO 20345 S3 CI/SRC, form B.