ESD Occupational Ankle Boots, Lace-up, Ronan XXF GTX O2 WR CI Mid
Leverandør: ELTEN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
976610/42 976610/43 976610/44 976610/45 976610/46 976610/47 976610/48 976610/35 976610/36 976610/37 976610/38 976610/39 976610/40 976610/41
ELTN976610/42EA 970 DKK
ELTN976610/42 ELTN976610/43 ELTN976610/44 ELTN976610/45 ELTN976610/46 ELTN976610/47 ELTN976610/48 ELTN976610/35 ELTN976610/36 ELTN976610/37 ELTN976610/38 ELTN976610/39 ELTN976610/40 ELTN976610/41
ESD Occupational Ankle Boots, Lace-up, Ronan XXF GTX O2 WR CI Mid
Sko og støvler
ESD work boot made from hydrophobised nubuck leather upper and a PU/PU sole (Wellmaxx Flex).
  • Full, padded bellows tongue
  • All weather Gore-Tex performance membrane
  • Full-length inlay sole ESD PRO black
  • Soft fleece ESD insole
Suitable for indoors and outdoors, areas where exposure to moisture is expected (O2), areas where there is no risk of falling heavy objects, areas where there is a risk of electrostatic discharge (ESDS/ESD).
Colour: Black
Certificeringer: EN ISO 20347 O2 CI/FO/WR/SR, form B.