ESD Occupational Shoes, Lacing Sytem, Riley XXF Boa® GTX Low O2 WR C
Leverandør: ELTEN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
972680/37 972680/38 972680/39 972680/40 972680/41 972680/42 972680/43 972680/44 972680/45 972680/46 972680/47 972680/48
ELTN972680/37EA 970 DKK
ELTN972680/37 ELTN972680/38 ELTN972680/39 ELTN972680/40 ELTN972680/41 ELTN972680/42 ELTN972680/43 ELTN972680/44 ELTN972680/45 ELTN972680/46 ELTN972680/47 ELTN972680/48
ESD Occupational Shoes, Lacing Sytem, Riley XXF Boa® GTX Low O2 WR C
Sko og støvler
ESD work shoe made from hydrophobised suede and hydrophobised textile upper and a PU/PU sole (Wellmaxx Flex).
  • Full, padded bellows tongue
  • All weather Gore-Tex performance membrane
  • Boa® fit lacing system
  • Full-length inlay sole ESD PRO grey
  • Soft fleece ESD insole
Suitable for use Indoors and outdoors, areas where exposure to moisture is expected (O2), a risk of electrostatic discharge (ESDS/ESD), areas where there is no risk of falling heavy objects.
Colour: Grey/lime
Certificeringer: EN ISO 20347 O2 WR/FO/SR/CI, form A.