ESD Safety Ankle Boots, Lacing Sytem, Mike Boa® Mid S3 S
Leverandør: ELTEN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
763831/44 763831/43 763831/45 763831/46 763831/47 763831/48 763831/40 763831/41 763831/42
ELTN763831/44EA 887 DKK
ELTN763831/44 ELTN763831/43 ELTN763831/45 ELTN763831/46 ELTN763831/47 ELTN763831/48 ELTN763831/40 ELTN763831/41 ELTN763831/42
ESD Safety Ankle Boots, Lacing Sytem, Mike Boa® Mid S3 S
Sko og støvler
ESD safety ankle boot made from hydrophobised microfibre and textile upper and a PU/PU sole (Crossworker).
  • Fabric lining for natural ventilation
  • Full, padded bellows tongue
  • Full-length inlay sole ESD PRO black
  • Boa® fit lacing system
  • Non-metallic, penetration-resistant midsole
  • Steel toe cap
  • PU scuff cap
Suitable for areas where there is a risk of electrostatic discharge (ESDS/ESD).
Colour: Black
Certificeringer: EN ISO 20345 S3S FO/SR/SC/LG, form B.