ESD Safety Ankle Boots, Lace-Up, Ian XXTP Pro Mid S3 S
Leverandør: ELTEN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
761321/38 761321/39 761321/40 761321/41 761321/48 761321/49 761321/35 761321/36 761321/37 761321/42 761321/43 761321/44 761321/45 761321/46 761321/47
ELTN761321/38EA 761 DKK
ELTN761321/38 ELTN761321/39 ELTN761321/40 ELTN761321/41 ELTN761321/48 ELTN761321/49 ELTN761321/35 ELTN761321/36 ELTN761321/37 ELTN761321/42 ELTN761321/43 ELTN761321/44 ELTN761321/45 ELTN761321/46 ELTN761321/47
ESD Safety Ankle Boots, Lace-Up, Ian XXTP Pro Mid S3 S
Sko og støvler
ESD safety ankle boots made from hydrophobised microfibre/hydrophobised textile material and a TPU/PU sole (Wellmaxx Trainers Power).
  • Fabric lining for natural ventilation
  • Full, padded bellows tongue
  • Full-length inlay sole ESD PRO black
  • Non-metallic, penetration-resistant midsole
  • Composite toe cap
  • TPU scuff cap
Suitable for areas where there is a risk of electrostatic discharge (ESDS/ESD).
Colour: Grey
Certificeringer: EN ISO 20345 S3S FO/SR/SC, form B.