ESD Safety Shoes, Lacing System, Matthew Pro Boa® GTX Low S3 CI Type 1
Leverandør: ELTEN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
7285501/46 7285501/47 7285501/48 1728501/36 1728501/37 1728501/38 1728501/39 7285501/40 7285501/41 7285501/42 7285501/43 7285501/44 7285501/45
ELTN7285501/46EA 970 DKK
ELTN7285501/46 ELTN7285501/47 ELTN7285501/48 ELTN1728501/36 ELTN1728501/37 ELTN1728501/38 ELTN1728501/39 ELTN7285501/40 ELTN7285501/41 ELTN7285501/42 ELTN7285501/43 ELTN7285501/44 ELTN7285501/45
ESD Safety Shoes, Lacing System, Matthew Pro Boa® GTX Low S3 CI Type 1
Sko og støvler
ESD safety shoe made from hydrophobised microfibre and hydrophobised textile Cordura® upper and a PU/PU sole (Ergo-Active 2). Type 1: For strong feet with short toes, wide ball and heel.
  • Gore-Tex Crosstech® membrane
  • Full, padded bellows tongue
  • Boa® fit lacing system
  • Semi-orthopaedic inlay sole ESD black
  • Non-metallic, penetration-resistant midsole
  • Steel toe cap
  • TPU scuff cap
Suitable for indoors and outdoors, areas with exposure to moisture (S2), risk of electrostatic discharge (ESDS/ESD) and areas where there is risk of penetration from pointed and sharp objects (S3).
Colour: Black
Shoe size for ladies: 36 to 39
Shoe size for men: 40 to 48
Certificeringer: EN ISO 20345 S3 CI/SRC, form A.