Anti-CD5 Mouse Monoclonal Antibody [clone: UCHT2] (PerCP (Peridinin-Chlorophyll Protein Complex))

Leverandør: AAT BIOQUEST
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

100521T0 100521T1 100521T2
AATB100521T0EA 2030 DKK
AATB100521T0 AATB100521T1 AATB100521T2
Anti-CD5 Mouse Monoclonal Antibody [clone: UCHT2] (PerCP (Peridinin-Chlorophyll Protein Complex))
Antistoffer
UCHT2 is an anti-human monoclonal antibody that is specific for the CD5 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Advarsel: For research use only.

Type: Primary
Antigen: CD5
Clonality: Monoclonal
Clone: UCHT2
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR