Anti-CD5 Mouse Monoclonal Antibody [clone: UCHT2] (PE (Phycoerythrin)/Cy5®)
Leverandør: AAT BIOQUEST
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
100521M0 100521M1 100521M2
AATB100521M0EA 2380 DKK
AATB100521M0 AATB100521M1 AATB100521M2
Anti-CD5 Mouse Monoclonal Antibody [clone: UCHT2] (PE (Phycoerythrin)/Cy5®)
Antistoffer
UCHT2 is an anti-human monoclonal antibody that is specific for the CD5 antigen.
  • Particularly useful for flow cytometry
This antibody was purified through affinity chromatography.
Advarsel: For research use only.
Type: Primary
Antigen: CD5
Clonality: Monoclonal
Clone: UCHT2
Conjugation: PE (Phycoerythrin)/Cy5®
Epitope:
Host: Mouse
Isotype:
Reactivity: Human