Anti-CD14 Mouse Monoclonal Antibody [clone: 61D3] (iFluor® A7)

Leverandør: AAT BIOQUEST
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

101410S0 101410S1
AATB101410S0EA 2770 DKK
AATB101410S0 AATB101410S1
Anti-CD14 Mouse Monoclonal Antibody [clone: 61D3] (iFluor® A7)
Antistoffer
61D3 is an anti-human monoclonal antibody that targets the CD14 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Advarsel: For research use only.

Type: Primary
Antigen: CD14
Clonality: Monoclonal
Clone: 61D3
Conjugation: iFluor® A7
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR