Anti-CD5 Mouse Monoclonal Antibody [clone: L17F12] (iFluor® 546)
Leverandør: AAT BIOQUEST
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
10051080 10051081
AATB10051080EA 2770 DKK
AATB10051080 AATB10051081
Anti-CD5 Mouse Monoclonal Antibody [clone: L17F12] (iFluor® 546)
Antistoffer
L17F12 is an anti-human monoclonal antibody that is specific for the CD5 antigen.
  • Particularly useful for flow cytometry
This antibody was purified through affinity chromatography.
Advarsel: For research use only.
Type: Primary
Antigen: CD5
Clonality: Monoclonal
Clone: L17F12
Conjugation: iFluor® 546
Epitope:
Host: Mouse
Isotype:
Reactivity: Human