Anti-CD19 Mouse Monoclonal Antibody [clone: SJ25C1] (PerCP/Cy5.5)

Leverandør: AAT BIOQUEST
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

101911U0 101911U1 101911U2
AATB101911U0EA 2380 DKK
AATB101911U0 AATB101911U1 AATB101911U2
Anti-CD19 Mouse Monoclonal Antibody [clone: SJ25C1] (PerCP/Cy5.5)
Antistoffer
SJ25C1 is an anti-human monoclonal antibody that forms an immune complex with the CD19 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Advarsel: For research use only.

Type: Primary
Antigen: CD19 antigen
Clonality: Monoclonal
Clone: SJ25C1
Conjugation: PerCP/Cy5.5
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR