Anti-CD5 Mouse Monoclonal Antibody [clone: L17F12] (PE/Cy5)

Leverandør: AAT BIOQUEST
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

100511M0 100511M1 100511M2
AATB100511M0EA 2380 DKK
AATB100511M0 AATB100511M1 AATB100511M2
Anti-CD5 Mouse Monoclonal Antibody [clone: L17F12] (PE/Cy5)
Antistoffer
L17F12 is an anti-human monoclonal antibody that is specific for the CD5 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Advarsel: For research use only.

Type: Primary
Antigen: CD5 antigen
Clonality: Monoclonal
Clone: L17F12
Conjugation: PE/Cy5
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR